siRNA Details
Gene Symbol: DAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005785.2 | NP_001005785.1 | 2, 3 | 445 | ||
NM_001005786.2 | NP_001005786.2 | 2, 3 | 445 | |||
NM_020363.3 | NP_065096.2 | 2, 3 | 445 | |||
GenBank mRNA | AF248480.1 | - | 2, 3 | 357 | ||
AF414184.1 | - | 2, 3 | 427 | |||
AK302398.1 | - | 2, 3 | 368 | |||
AK302690.1 | - | 1, 2 | 298 | |||
BC113006.1 | - | 2, 3 | 253 | 40035862 | ||
U21663.1 | - | 2, 3 | 369 |
Gene Symbol: DAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004081.5 | NP_004072.3 | 2, 3 | 453 | ||
XM_011531482.2 | XP_011529784.1 | 3 | 491 | |||
XM_011531483.1 | XP_011529785.1 | 2 | 113 | |||
GenBank mRNA | AF271087.1 | - | 2, 3 | 295 | ||
AF271088.1 | - | 4, 5 | 423 | |||
BC114927.1 | - | 2, 3 | 284 | 40028543 | ||
DB461291.1 | - | 2, 3 | 453 |
Gene Symbol: DAZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020364.3 | NP_065097.2 | 2, 3 | 468 | ||
GenBank mRNA | AF248481.1 | - | 2, 3 | 349 | ||
BC113005.1 | - | 2, 3 | 253 | 40035860 |
Gene Symbol: DAZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005375.2 | NP_001005375.1 | 2, 3 | 468 | ||
NM_020420.3 | NP_065153.1 | 2, 3 | 468 | |||
XM_005262602.1 | XP_005262659.1 | 2, 3 | 407 | |||
XM_011531509.2 | XP_011529811.1 | 3 | 494 | |||
XM_011531510.1 | XP_011529812.1 | 2 | 113 | |||
GenBank mRNA | AF248482.1 | - | 2, 3 | 357 | ||
AF248483.1 | - | 1, 2 | 158 | |||
BC047480.1 | - | 2, 3 | 373 | 5297459 | ||
BC047617.1 | - | 2, 3 | 369 | 4836748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
