siRNA Details
Gene Symbol: DAZ3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020364.3 | NP_065097.2 | 19 | 3297 |
Gene Symbol: DAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004081.5 | NP_004072.3 | 28 | 4200 | ||
XM_011531482.2 | XP_011529784.1 | 28 | 4238 | |||
XM_011531483.1 | XP_011529785.1 | 22 | 3365 | |||
GenBank mRNA | U66077.1 | - | 2 | 1673 |
Gene Symbol: DAZ2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005785.2 | NP_001005785.1 | 23 | 3562 | ||
NM_001005786.2 | NP_001005786.2 | 16 | 3058 | |||
NM_020363.3 | NP_065096.2 | 24 | 3634 | |||
GenBank mRNA | AF414184.1 | - | 23 | 3544 | ||
AK302398.1 | - | 26 | 3701 | |||
AK302690.1 | - | 27 | 3703 |
Gene Symbol: DAZ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005375.2 | NP_001005375.1 | 23 | 3720 | ||
NM_020420.3 | NP_065153.1 | 17 | 3153 | |||
XM_005262602.1 | XP_005262659.1 | 29 | 4091 | |||
XM_011531509.2 | XP_011529811.1 | 29 | 4178 | |||
XM_011531510.1 | XP_011529812.1 | 23 | 3302 | |||
GenBank mRNA | AL833003.1 | - | 5 | 2031 | ||
BC047480.1 | - | 17 | 3058 | 5297459 | ||
BC047617.1 | - | 17 | 3054 | 4836748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
