siRNA Details
Gene Symbol: LOC441259 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC075816.1 | - | 2 | 126 |
Gene Symbol: PMS2P10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC047753.1 | - | 2 | 122 |
Gene Symbol: PMS2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003614.3 | 2 | 146 | |||
GenBank mRNA | DA215155.1 | - | 2 | 133 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027775.2 | 2 | 171 | |||
NR_027776.2 | 2 | 171 | ||||
NR_027777.2 | 2 | 171 | ||||
GenBank mRNA | AK309660.1 | - | 2 | 119 | ||
BC052588.1 | - | 2 | 174 | |||
BF984173.1 | - | 2 | 170 | |||
D38436.1 | - | 2 | 152 | |||
D38439.1 | - | 1 | 83 | |||
D38501.1 | - | 1 | 83 | |||
D38502.1 | - | 2 | 152 | |||
DW407867.1 | - | 2 | 188 |
Gene Symbol: PMS2P6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB017005.1 | - | 3 | 324 |
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 2 | 187 | |||
GenBank mRNA | AB017006.1 | - | 2 | 282 | ||
DB472687.1 | - | 2 | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
