siRNA Details
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164803.1 | NP_001158275.1 | 2 | 259 | ||
NM_002139.3 | NP_002130.2 | 2 | 259 | |||
NR_028476.1 | 2 | 259 | ||||
NR_028477.1 | 2 | 259 | ||||
GenBank mRNA | AK091520.1 | - | 2 | 205 | ||
AK096015.1 | - | 1 | 204 | |||
AK304573.1 | - | 2 | 204 | |||
AK304868.1 | - | 2 | 204 | |||
AK309911.1 | - | 2 | 214 | |||
AY464692.1 | - | 1 | 49 | |||
BC006550.2 | - | 2 | 195 | 3010123 | ||
BC007435.2 | - | 2 | 195 | 3010123 | ||
BM147410.1 | - | 2 | 259 | |||
BU587845.1 | - | 2 | 263 | |||
BX647131.1 | - | 2 | 202 | |||
DA925805.1 | - | 2 | 204 | |||
DC337568.1 | - | 2 | 204 | |||
Z23064.1 | - | 1 | 60 |
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162536.2 | NP_001156008.1 | 3 | 765 | ||
NM_019610.5 | NP_062556.2 | 2 | 665 | |||
GenBank mRNA | AL832554.1 | - | 2 | 517 | ||
BC012942.1 | - | 2 | 324 | 3875565 | ||
DA390224.1 | - | 2 | 481 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
