siRNA Details
Gene Symbol: KYAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008661.2 | NP_001008661.1 | 14 | 2082 | ||
NM_001008662.2 | NP_001008662.1 | 13 | 1982 | |||
GenBank mRNA | AK057176.1 | - | 13 | 1798 | ||
AY028624.1 | - | 14 | 1902 | |||
BC000819.1 | - | 11 | 1455 | 3453829 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
