siRNA Details
Gene Symbol: ZNF844 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136501.2 | NP_001129973.1 | 4 | 812 | ||
NR_134326.1 | 3 | 751 | ||||
XM_017026664.1 | XP_016882153.1 | 3 | 658 | |||
GenBank mRNA | AK027865.1 | - | 1 | 98 | ||
AK098117.1 | - | 4 | 780 | |||
AK299732.1 | - | 4 | 812 | |||
AL832297.1 | - | 4 | 798 | |||
BC125185.1 | - | 3 | 640 | |||
BC125186.1 | - | 4 | 698 | 40122307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
