siRNA Details
Gene Symbol: SPACA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205856.3 | NP_995328.2 | 2 | 214 | ||
XM_017029512.1 | XP_016885001.1 | 1 | 829 | |||
GenBank mRNA | AF217622.1 | - | 1 | 70 | ||
AY358844.1 | - | 2 | 214 | |||
BX279755.1 | - | 1 | 63 |
Gene Symbol: SPACA5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079900.1 | NP_001073369.1 | 1 | 77 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
