siRNA Details
Gene Symbol: PDE4DIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002811.2 | NP_001002811.2 | 19 | 5356 | ||
NM_001002812.2 | NP_001002812.2 | 23 | 4512 | |||
GenBank mRNA | AB007946.1 | - | 19 | 5356 | ||
AL831815.2 | - | 23 | 4355 | |||
AL832024.2 | - | 22 | 4601 | |||
BC078660.1 | - | 19 | 4862 | |||
BC152439.1 | - | 19 | 5356 |
Gene Symbol: LOC100996724 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK001619.1 | - | 12 | 3082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
