siRNA Details
Gene Symbol: LOC653513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003021.1 | XP_016858510.1 | 5 | 906 | ||
XM_017003022.1 | XP_016858511.1 | 5 | 906 | |||
XM_017003023.1 | XP_016858512.1 | 4 | 795 | |||
XM_017003024.1 | XP_016858513.1 | 5 | 906 | |||
XM_017003025.1 | XP_016858514.1 | 5 | 906 | |||
XM_017003026.1 | XP_016858515.1 | 5 | 906 | |||
XM_017003027.1 | XP_016858516.1 | 5 | 741 | |||
XM_017003028.1 | XP_016858517.1 | 4 | 795 | |||
XM_017003029.1 | XP_016858518.1 | 4 | 795 | |||
XM_017003030.1 | XP_016858519.1 | 4 | 637 | |||
XM_017003031.1 | XP_016858520.1 | 5 | 741 | |||
XM_017003032.1 | XP_016858521.1 | 5 | 741 | |||
XM_017003033.1 | XP_016858522.1 | 5 | 741 | |||
XM_017003036.1 | XP_016858525.1 | 4 | 637 | |||
XM_017003037.1 | XP_016858526.1 | 4 | 637 | |||
XM_017003038.1 | XP_016858527.1 | 2 | 1516 | |||
XM_017003040.1 | XP_016858529.1 | 3 | 249 | |||
GenBank mRNA | EL953210.1 | - | 1 | 74 |

More Information
Other Species Targeted

Gene Ontology Categories:
