siRNA Details
Gene Symbol: GNG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017998.3 | NP_001017998.1 | 2 | 255 | ||
NM_001198664.1 | NP_001185593.1 | 2 | 255 | |||
GenBank mRNA | AF493877.1 | - | 2 | 191 | ||
BC010384.1 | - | 2 | 217 | |||
BC015206.2 | - | 2 | 201 | |||
BC016319.1 | - | 2 | 213 | |||
BC072671.1 | - | 2 | 206 | 5768882 | ||
BC107574.1 | - | 2 | 222 | 4050245 | ||
BG708408.1 | - | 2 | 260 | |||
DB504443.1 | - | 2 | 255 | |||
U31383.1 | - | 2 | 214 |
Gene Symbol: DNAJC25-GNG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004125.3 | NP_004116.2 | 2 | 502 | ||
GenBank mRNA | BC015391.1 | - | 2 | 468 | 4425229 | |
BT007068.1 | - | 2 | 446 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
