siRNA Details
Gene Symbol: SFTPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098668.3 | NP_001092138.1 | 6 | 683 | ||
NM_001320813.1 | NP_001307742.1 | 6 | 686 | |||
NM_001320814.1 | NP_001307743.1 | 5 | 752 | |||
XM_005270128.3 | XP_005270185.1 | 6 | 664 | |||
XM_005270132.3 | XP_005270189.1 | 5 | 651 | |||
XM_011540124.1 | XP_011538426.1 | 6 | 779 | |||
XM_011540125.1 | XP_011538427.1 | 6 | 722 | |||
XM_017016608.1 | XP_016872097.1 | 5 | 575 | |||
GenBank mRNA | AK225599.1 | - | 6 | 629 | ||
AK298041.1 | - | 6 | 536 | |||
AK298074.1 | - | 5 | 355 | |||
AK298114.1 | - | 6 | 482 | |||
AK313179.1 | - | 6 | 632 | |||
AK316192.1 | - | 6 | 562 | |||
BC111571.1 | - | 6 | 608 | 40069329 | ||
BC139727.1 | - | 4 | 555 | 40024708 | ||
BC157866.1 | - | 6 | 622 | 9057085 | ||
BC157890.1 | - | 6 | 622 | 8862711 | ||
DC358710.1 | - | 5 | 416 | |||
HQ021421.1 | - | 6 | 632 | |||
HQ021422.2 | - | 6 | 632 | |||
HQ021423.2 | - | 6 | 632 | |||
HQ021424.1 | - | 6 | 632 | |||
HQ021425.2 | - | 6 | 632 | |||
HQ021426.1 | - | 6 | 629 | |||
HQ021427.1 | - | 6 | 629 | |||
HQ021428.2 | - | 6 | 629 | |||
HQ021429.2 | - | 6 | 629 | |||
HQ021430.1 | - | 6 | 629 | |||
HQ021431.2 | - | 6 | 629 | |||
HQ021432.1 | - | 6 | 632 | |||
K03475.1 | - | 6 | 620 |
Gene Symbol: SFTPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001093770.2 | NP_001087239.2 | 6 | 659 | ||
NM_001164644.1 | NP_001158116.1 | 6 | 629 | |||
NM_001164645.1 | NP_001158117.1 | 6 | 515 | |||
NM_001164646.1 | NP_001158118.1 | 6 | 523 | |||
NM_001164647.1 | NP_001158119.1 | 5 | 599 | |||
NM_005411.4 | NP_005402.3 | 6 | 670 | |||
XM_005270062.4 | XP_005270119.1 | 5 | 659 | |||
XM_006717953.2 | XP_006718016.1 | 6 | 652 | |||
GenBank mRNA | AK290703.1 | - | 6 | 661 | ||
AK298002.1 | - | 6 | 515 | |||
AK298029.1 | - | 5 | 548 | |||
AK298034.1 | - | 5 | 406 | |||
AK298092.1 | - | 4 | 401 | |||
AK309518.1 | - | 5 | 671 | |||
AK316196.1 | - | 5 | 323 | |||
BC026229.1 | - | 1 | 450 | |||
BC111570.1 | - | 5 | 578 | 40069324 | ||
BC171875.1 | - | 5 | 589 | 9054529 | ||
HQ021433.2 | - | 5 | 599 | |||
HQ021434.1 | - | 5 | 599 | |||
HQ021435.1 | - | 5 | 599 | |||
HQ021436.1 | - | 5 | 599 | |||
HQ021437.1 | - | 6 | 670 | |||
HQ021438.1 | - | 6 | 670 | |||
HQ021439.1 | - | 6 | 670 | |||
HQ021440.1 | - | 6 | 659 | |||
HQ021441.1 | - | 6 | 659 | |||
HQ021442.1 | - | 6 | 659 | |||
JX502764.1 | - | 6 | 670 | |||
JX502765.1 | - | 6 | 659 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
