siRNA Details
Gene Symbol: ABCC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079528.3 | NP_001072996.1 | 2 | 659 | ||
XM_011522481.2 | XP_011520783.1 | 1 | 48 | |||
GenBank mRNA | AY078405.1 | - | 2 | 668 | ||
BC050733.1 | - | 2 | 644 |
Gene Symbol: ABCC6P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023387.1 | 2 | 655 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
