siRNA Details
Gene Symbol: ABCC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171.5 | NP_001162.4 | 4 | 397 | ||
XM_011522479.2 | XP_011520781.1 | 4 | 541 | |||
XM_011522480.1 | XP_011520782.1 | 4 | 345 | |||
XM_011522481.2 | XP_011520783.1 | 4 | 544 | |||
XM_011522482.2 | XP_011520784.1 | 4 | 538 | |||
XM_017023212.1 | XP_016878701.1 | 4 | 541 | |||
XM_017023213.1 | XP_016878702.1 | 4 | 431 | |||
XM_017023214.1 | XP_016878703.1 | 4 | 542 | |||
GenBank mRNA | AF076622.1 | - | 4 | 383 | ||
AF168791.1 | - | 4 | 360 | |||
AK298295.1 | - | 4 | 456 | |||
AM711638.1 | - | 4 | 360 | |||
AM774324.1 | - | 4 | 360 | |||
BC131732.1 | - | 4 | 383 | 40124570 | ||
DC361874.1 | - | 4 | 456 |
Gene Symbol: ABCC6P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003569.1 | 4 | 389 | |||
GenBank mRNA | AK298443.1 | - | 5 | 565 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
