siRNA Details
Gene Symbol: EIF4EBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003732.2 | NP_003723.1 | 3 | 491 | ||
GenBank mRNA | AF038869.1 | - | 3 | 492 | ||
BC010881.1 | - | 3 | 487 | |||
BC073751.1 | - | 3 | 466 | 5814294 |
Gene Symbol: ANKHD1-EIF4EBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020690.5 | NP_065741.3 | 36 | 8149 | ||
GenBank mRNA | AB029008.1 | - | 7 | 5255 | ||
AF521883.1 | - | 36 | 8055 | |||
AK125734.1 | - | 9 | 2113 | |||
BC144623.1 | - | 36 | 8234 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
