siRNA Details
Gene Symbol: FOXD4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012184.4 | NP_036316.1 | 1 | 1624 | ||
GenBank mRNA | AK122950.1 | - | 1 | 1624 | ||
AK125398.1 | - | 1 | 2043 | |||
AY344639.1 | - | 1 | 1536 | |||
BC136428.1 | - | 1 | 1616 | 9020417 |
Gene Symbol: FOXD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207305.4 | NP_997188.2 | 1 | 1739 | ||
GenBank mRNA | AY344640.1 | - | 1 | 1527 | ||
BC089432.1 | - | 1 | 1742 | 30555309 | ||
BC136570.1 | - | 1 | 1474 | 9020560 | ||
BC136571.1 | - | 1 | 1474 | 9020561 |
Gene Symbol: FOXD4L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199135.4 | NP_954586.4 | 1 | 1591 | ||
GenBank mRNA | AY344641.1 | - | 1 | 1591 | ||
AY344642.1 | - | 1 | 1591 |
Gene Symbol: FOXD4L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085476.1 | NP_001078945.1 | 1 | 2098 | ||
GenBank mRNA | BC137365.1 | - | 1 | 1586 | 9021363 | |
BC137366.1 | - | 1 | 1586 | 9021364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
