siRNA Details
Gene Symbol: DEFB119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271209.1 | NP_001258138.1 | 1 | 177 | ||
NM_153289.3 | NP_695021.2 | 1 | 177 | |||
NM_153323.4 | NP_697018.1 | 1 | 177 | |||
NR_073151.1 | 1 | 177 | ||||
NR_073152.1 | 1 | 177 | ||||
NR_073153.1 | 1 | 177 | ||||
NR_126440.1 | 1 | 177 | ||||
GenBank mRNA | AF479698.1 | - | 11 | |||
AF479699.1 | - | 11 | ||||
AL732301.1 | - | 1 | 97 | |||
AY122472.1 | - | 1 | 131 | |||
AY122473.1 | - | 11 | ||||
AY358860.1 | - | 1 | 103 | |||
AY501001.1 | - | 1 | 57 | |||
BC062212.1 | - | 1 | 122 | 6621528 | ||
BI460707.1 | - | 1 | 122 | |||
BU561156.1 | - | 1 | 74 | |||
HY035342.1 | - | 1 | 177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
