siRNA Details
Gene Symbol: RPL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098577.2 | NP_001092047.1 | 5 | 980 | ||
GenBank mRNA | AK297483.1 | - | 5 | 916 | ||
CN269915.1 | - | 2 | 577 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
