siRNA Details
Gene Symbol: RPL31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000993.4 | NP_000984.1 | 4 | 390 | ||
NM_001098577.2 | NP_001092047.1 | 4 | 390 | |||
NM_001099693.1 | NP_001093163.1 | 4 | 390 | |||
GenBank mRNA | AK297167.1 | - | 4 | 327 | ||
AK297247.1 | - | 4 | 327 | |||
AK297483.1 | - | 4 | 326 | |||
AK311775.1 | - | 4 | 327 | |||
BC017343.2 | - | 3 | 305 | 4895396 | ||
BC070210.1 | - | 4 | 326 | 4714258 | ||
BC070373.1 | - | 1 | 326 | 4666695 | ||
BE564858.1 | - | 3 | 308 | |||
BG031488.1 | - | 3 | 307 | |||
BG223187.1 | - | 1 | 344 | |||
CD108396.1 | - | 1 | 298 | |||
CD247800.1 | - | 4 | 359 | |||
CF124259.1 | - | 4 | 384 | |||
CN269912.1 | - | 3 | 270 | |||
X15940.1 | - | 3 | 310 | |||
X69181.1 | - | 3 | 291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
