siRNA Details
Gene Symbol: KRTAP10-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198691.2 | NP_941964.2 | 1 | 470 | ||
GenBank mRNA | AB076347.1 | - | 1 | 470 | ||
BC120959.2 | - | 1 | 464 | 40115271 | ||
BC120960.1 | - | 1 | 464 | 40115274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
