siRNA Details
Gene Symbol: CGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319065.1 | NP_001305994.1 | 1 | 123 | ||
NM_033378.1 | NP_203696.2 | 1 | 162 | |||
GenBank mRNA | BC069367.1 | - | 1 | 57 | 7262156 | |
BC107143.1 | - | 1 | 123 |
Gene Symbol: CGB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033377.1 | NP_203695.2 | 1 | 162 | ||
GenBank mRNA | BC107144.1 | - | 1 | 123 | ||
BC126460.1 | - | 1 | 44 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
