siRNA Details
Gene Symbol: VCX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016378.3 | NP_057462.2 | 3 | 633 | ||
GenBank mRNA | AF159127.1 | - | 2 | 381 | ||
AF167079.1 | - | 3 | 663 | |||
BC096715.1 | - | 2 | 381 | 40002645 | ||
BC098101.1 | - | 2 | 381 | 40002641 | ||
BC098113.1 | - | 2 | 381 | 40014419 | ||
BC098163.1 | - | 2 | 381 | 40014421 | ||
BC104153.2 | - | 2 | 381 | 40029400 | ||
BC104167.1 | - | 2 | 472 | 40030354 | ||
BC104168.1 | - | 2 | 472 | 40030355 | ||
BC134350.1 | - | 2 | 427 | 40114437 |
Gene Symbol: VCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013452.2 | NP_038480.2 | 3 | 575 | ||
XM_011545490.2 | XP_011543792.1 | 2 | 467 | |||
GenBank mRNA | AF159129.1 | - | 2 | 382 | ||
AF167081.2 | - | 3 | 575 | |||
BC098123.1 | - | 2 | 381 | 40002642 | ||
BC098149.1 | - | 2 | 381 | 40002644 | ||
BC126903.1 | - | 2 | 472 | 40030361 |
Gene Symbol: VCX3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016379.3 | NP_057463.2 | 3 | 663 | ||
XM_006724494.3 | XP_006724557.1 | 2, 3 | 447 | |||
GenBank mRNA | AF159128.1 | - | 2 | 362 | ||
AF167078.1 | - | 3 | 663 | |||
BC039722.1 | - | 2 | 374 | |||
BC126902.1 | - | 2, 3 | 472 | 40030357 |
Gene Symbol: VCX3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001888.3 | NP_001001888.3 | 3 | 663 | ||
GenBank mRNA | AF167080.2 | - | 3 | 663 | ||
BC098143.1 | - | 3, 4 | 435 | 40014420 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
