siRNA Details
Gene Symbol: AQP12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102467.1 | NP_001095937.1 | 3 | 1051 | ||
XM_011511671.2 | XP_011509973.1 | 9 | 1764 | |||
XM_011511672.2 | XP_011509974.1 | 8 | 1704 | |||
XM_011511673.2 | XP_011509975.1 | 9 | 1695 | |||
XM_011511675.2 | XP_011509977.1 | 7 | 1626 | |||
XM_011511676.2 | XP_011509978.1 | 6 | 1436 | |||
XM_017004736.1 | XP_016860225.1 | 8 | 1707 | |||
XM_017004737.1 | XP_016860226.1 | 5 | 1269 | |||
GenBank mRNA | BC041460.1 | - | 5 | 2950 | ||
BC139748.1 | - | 3 | 1051 | 40115528 | ||
BC157895.1 | - | 3 | 1044 | 8862714 | ||
BC171912.1 | - | 3 | 1044 | 9054566 |
Gene Symbol: AQP12A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198998.2 | NP_945349.1 | 4 | 1021 | ||
GenBank mRNA | AB040748.1 | - | 4 | 1021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
