siRNA Details
Gene Symbol: KRTAP10-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198696.2 | NP_941969.2 | 1 | 52 | ||
GenBank mRNA | AB076350.1 | - | 1 | 52 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
