siRNA Details
Gene Symbol: KALRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024660.4 | NP_001019831.2 | 50 | 7452 | ||
NM_001322993.1 | NP_001309922.1 | 17 | 2364 | |||
NM_007064.4 | NP_008995.2 | 17 | 2367 | |||
XM_006713810.3 | XP_006713873.1 | 50 | 7697 | |||
XM_006713811.3 | XP_006713874.1 | 50 | 7670 | |||
XM_006713812.3 | XP_006713875.1 | 50 | 7658 | |||
XM_006713813.3 | XP_006713876.1 | 50 | 7631 | |||
XM_006713814.3 | XP_006713877.1 | 49 | 7601 | |||
XM_011513279.2 | XP_011511581.1 | 50 | 7694 | |||
XM_011513280.2 | XP_011511582.1 | 50 | 7695 | |||
XM_011513281.2 | XP_011511583.1 | 50 | 30319 | |||
XM_017007429.1 | XP_016862918.1 | 50 | 7697 | |||
XM_017007430.1 | XP_016862919.1 | 49 | 7598 | |||
GenBank mRNA | AB011422.1 | - | 17 | 2321 | ||
BM930007.1 | - | 3 | 419 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_177357.
Gene Ontology Categories:
