siRNA Details
Gene Symbol: FOXP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244808.1 | NP_001231737.1 | 21 | 4587 | ||
NM_001244810.1 | NP_001231739.1 | 21 | 4535 | |||
NM_001244812.1 | NP_001231741.1 | 16 | 3887 | |||
NM_001244813.1 | NP_001231742.1 | 15 | 3779 | |||
NM_001244814.1 | NP_001231743.1 | 17 | 4284 | |||
NM_001244815.1 | NP_001231744.1 | 16 | 4225 | |||
NM_001244816.1 | NP_001231745.1 | 20 | 4392 | |||
NM_032682.5 | NP_116071.2 | 21 | 4487 | |||
XM_006713102.2 | XP_006713165.1 | 20 | 4448 | |||
XM_011533584.2 | XP_011531886.1 | 21 | 4558 | |||
XM_011533585.2 | XP_011531887.1 | 20 | 4410 | |||
XM_017006165.1 | XP_016861654.1 | 20 | 4329 | |||
GenBank mRNA | AK026898.1 | - | 1 | 1287 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
