siRNA Details
Gene Symbol: CGB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033142.1 | NP_149133.1 | 1 | 331 | ||
XM_017027502.1 | XP_016882991.1 | 4 | 1678 | |||
GenBank mRNA | AK125108.1 | - | 3 | 2838 |
Gene Symbol: CGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000737.3 | NP_000728.1 | 1 | 370 | ||
GenBank mRNA | AK291552.1 | - | 1 | 328 | ||
BC022796.1 | - | 1 | 308 | 4811294 | ||
BC106059.1 | - | 1 | 316 | 4992804 | ||
BX334905.2 | - | 1 | 372 | |||
K03181.1 | - | 1 | 158 |
Gene Symbol: CGB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033043.1 | NP_149032.1 | 1 | 331 | ||
GenBank mRNA | BC006290.2 | - | 1 | 292 | 3960125 | |
BC051378.1 | - | 1 | 292 | 3956670 | ||
BC103970.1 | - | 1 | 62 | 40014677 | ||
BC103971.1 | - | 1 | 58 | 40014679 |
Gene Symbol: CGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC103969.1 | - | 1 | 58 | 40014676 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
