siRNA Details
Gene Symbol: WWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270453.1 | NP_001257382.1 | 14 | 1540 | ||
NM_001270454.1 | NP_001257383.1 | 17 | 1850 | |||
NM_007014.4 | NP_008945.2 | 18 | 1903 | |||
NM_199424.2 | NP_955456.1 | 7 | 995 | |||
XM_011522823.2 | XP_011521125.1 | 19 | 1933 | |||
XM_011522825.1 | XP_011521127.1 | 17 | 1814 | |||
XM_011522826.2 | XP_011521128.1 | 14 | 1704 | |||
XM_017022879.1 | XP_016878368.1 | 19 | 2007 | |||
XM_017022880.1 | XP_016878369.1 | 19 | 1939 | |||
XM_017022881.1 | XP_016878370.1 | 19 | 1983 | |||
GenBank mRNA | AK126332.1 | - | 7 | 980 | ||
AK295070.1 | - | 10 | 895 | |||
AK295701.1 | - | 15 | 1599 | |||
AK300266.1 | - | 14 | 1540 | |||
AK312792.1 | - | 17 | 1830 | |||
AK316097.1 | - | 8 | 849 | |||
BC013645.1 | - | 16 | 1737 | 3453212 | ||
BC064531.1 | - | 17 | 1789 | 5588092 | ||
BU167468.1 | - | 396 | ||||
JN712744.1 | - | 17 | 1820 | |||
U96114.1 | - | 18 | 1835 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
