siRNA Details
Gene Symbol: WWP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270453.1 | NP_001257382.1 | 2 | 187 | ||
NM_001270454.1 | NP_001257383.1 | 5 | 497 | |||
NM_001270455.1 | NP_001257384.1 | 5 | 497 | |||
NM_007014.4 | NP_008945.2 | 6 | 550 | |||
XM_011522823.2 | XP_011521125.1 | 7 | 580 | |||
XM_011522825.1 | XP_011521127.1 | 5 | 461 | |||
XM_011522826.2 | XP_011521128.1 | 2 | 351 | |||
XM_017022879.1 | XP_016878368.1 | 7 | 654 | |||
XM_017022880.1 | XP_016878369.1 | 7 | 586 | |||
XM_017022881.1 | XP_016878370.1 | 7 | 630 | |||
GenBank mRNA | AJ001982.1 | - | 5 | 478 | ||
AK295701.1 | - | 3 | 246 | |||
AK300266.1 | - | 2 | 187 | |||
AK312792.1 | - | 5 | 477 | |||
BC000108.2 | - | 4 | 399 | |||
BC013645.1 | - | 4 | 384 | 3453212 | ||
BC064531.1 | - | 5 | 436 | 5588092 | ||
DC402912.1 | - | 5 | 497 | |||
JN712744.1 | - | 5 | 467 | |||
U96114.1 | - | 6 | 482 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
