siRNA Details
Gene Symbol: H2AFV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012412.4 | NP_036544.1 | 5 | 507 | ||
NM_201436.2 | NP_958844.1 | 4 | 429 | |||
NM_201516.2 | NP_958924.1 | 4 | 377 | |||
NM_201517.2 | NP_958925.1 | 4 | 393 | |||
GenBank mRNA | AB209001.1 | - | 5 | 418 | ||
AF081192.1 | - | 5 | 372 | |||
AK129989.1 | - | 5 | 427 | |||
AL520785.1 | - | 2 | 155 | |||
AL520811.3 | - | 5 | 408 | |||
BC000098.2 | - | 5 | 434 | 3505930 | ||
BC004274.1 | - | 4 | 359 | 3615320 | ||
BC014885.2 | - | 5 | 403 | 3907411 | ||
BC070169.1 | - | 5 | 434 | 6458566 | ||
BG754026.1 | - | 5 | 482 | |||
BG773256.1 | - | 5 | 510 | |||
BI257891.1 | - | 5 | 465 | |||
BM453663.1 | - | 5 | 377 | |||
BX644511.1 | - | 1 | 33 | |||
CB148943.1 | - | 4 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
