siRNA Details
Gene Symbol: GOLGA6L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001413.3 | NP_001001413.3 | 3, 4 | 477 | ||
XM_017022085.1 | XP_016877574.1 | 3, 4 | 494 | |||
GenBank mRNA | AK097517.1 | - | 3, 4 | 459 | ||
BC127816.1 | - | 3, 4 | 261 |
Gene Symbol: GOLGA6L22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271664.2 | NP_001258593.2 | 4, 5 | 419 | ||
XM_011543882.1 | XP_011542184.1 | 3, 4 | 459 |
Gene Symbol: GOLGA6L6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145004.2 | NP_001138476.2 | 3, 4 | 441 |
Gene Symbol: LOC100132202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017022770.1 | XP_016878259.1 | 3, 4 | 281 |
Gene Symbol: LOC102723623 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006720795.3 | XP_006720858.1 | 3, 4 | 475 | ||
XM_017022778.1 | XP_016878267.1 | 3, 4 | 479 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
