siRNA Details
Gene Symbol: SAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000331.5 | NP_000322.2 | 3 | 371 | ||
NM_001178006.2 | NP_001171477.1 | 4 | 285 | |||
NM_199161.4 | NP_954630.1 | 3 | 224 | |||
GenBank mRNA | BC007022.1 | - | 3 | 185 | 4071612 | |
BC105796.1 | - | 4 | 250 | 4722684 | ||
BG533276.1 | - | 333 | ||||
BG562565.1 | - | 3 | 224 | |||
BG565078.1 | - | 3 | 333 | |||
BG567902.1 | - | 276 | ||||
BQ691948.1 | - | 2 | 87 | |||
BX102362.1 | - | 3 | 291 | |||
CD102084.1 | - | 3 | 188 | |||
CR542241.1 | - | 2 | 148 | |||
M10906.1 | - | 2 | 148 | |||
M23698.1 | - | 2 | 148 | |||
X51443.1 | - | 2 | 127 |
Gene Symbol: SAA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127380.2 | NP_001120852.1 | 3 | 224 | ||
NM_030754.4 | NP_110381.2 | 3 | 224 | |||
GenBank mRNA | AK307163.1 | - | 3 | 184 | ||
BC020795.1 | - | 3 | 185 | 4767650 | ||
M23699.1 | - | 2 | 148 | |||
M23700.1 | - | 2 | 148 | |||
M26152.1 | - | 3 | 171 | |||
X51444.1 | - | 3 | 208 | |||
X51445.1 | - | 2 | 139 |
Gene Symbol: SAA2-SAA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199744.1 | NP_001186673.1 | 3 | 224 | ||
GenBank mRNA | BG616641.1 | - | 3 | 186 | ||
BX117371.1 | - | 3 | 168 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
