siRNA Details
Gene Symbol: CGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033183.2 | NP_149439.1 | 1 | 373 | ||
GenBank mRNA | BC030994.1 | - | 1 | 368 | 4605021 | |
BC103969.1 | - | 1 | 93 | 40014676 | ||
BC128603.1 | - | 1 | 93 | 40002893 | ||
BX118262.1 | - | 1 | 373 | |||
BX380841.2 | - | 1 | 353 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
