siRNA Details
Gene Symbol: CGB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033183.2 | NP_149439.1 | 1 | 338 | ||
GenBank mRNA | BC030994.1 | - | 1 | 333 | 4605021 | |
BC128603.1 | - | 1 | 58 | 40002893 | ||
BX118262.1 | - | 1 | 338 | |||
BX380841.2 | - | 1 | 318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
