siRNA Details
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021967.2 | NP_068802.1 | 3 | 710 | ||
GenBank mRNA | AF073519.1 | - | 3 | 686 | ||
BC035932.1 | - | 3 | 639 | 4701505 |
Gene Symbol: SERF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022978.2 | NP_075267.1 | 3 | 704 | ||
GenBank mRNA | BC103687.1 | - | 3 | 524 | 40000408 | |
BC104738.1 | - | 3 | 524 | 40000411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
