siRNA Details
Gene Symbol: TOMM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001790.2 | NP_001001790.1 | 1, 2 | 237 | ||
GenBank mRNA | AB435233.1 | - | 1, 2 | 130 | ||
BC034247.1 | - | 1, 2 | 169 | 3861303 | ||
DB451927.1 | - | 1, 2 | 240 |
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021967.2 | NP_068802.1 | 3 | 1556 | ||
GenBank mRNA | AF073519.1 | - | 3 | 1533 | ||
BC035932.1 | - | 3 | 1483 | 4701505 |
Gene Symbol: SERF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022978.2 | NP_075267.1 | 3 | 1550 | ||
GenBank mRNA | BM686659.1 | - | 1 | 219 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
