siRNA Details
Gene Symbol: SERF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022978.2 | NP_075267.1 | 3 | 522 | ||
GenBank mRNA | BC101970.1 | - | 3 | 342 | 40000410 | |
BC103687.1 | - | 3 | 342 | 40000408 | ||
BC104738.1 | - | 3 | 342 | 40000411 | ||
BC143255.1 | - | 3 | 340 | 9051757 | ||
BC143260.1 | - | 3 | 344 | 9051762 | ||
BC171749.1 | - | 3 | 344 | 9053250 |
Gene Symbol: SERF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021967.2 | NP_068802.1 | 3 | 528 | ||
GenBank mRNA | AF073519.1 | - | 3 | 504 | ||
BC035932.1 | - | 3 | 457 | 4701505 | ||
BC093880.1 | - | 3 | 344 | 7939725 | ||
BC093882.1 | - | 3 | 340 | 7939727 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
