siRNA Details
Gene Symbol: TRIM51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032681.3 | NP_116070.2 | 7 | 1195 | ||
GenBank mRNA | BC005014.1 | - | 5 | 835 | 3635781 | |
JF968454.1 | - | 4 | 437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
