siRNA Details
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 4 | 724 | ||
GenBank mRNA | AK296338.1 | - | 4 | 565 | ||
AK303264.1 | - | 1 | 37 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 6 | 638 | ||
XM_017023514.1 | XP_016879003.1 | 6 | 927 | |||
XM_017023515.1 | XP_016879004.1 | 7 | 946 | |||
XM_017023516.1 | XP_016879005.1 | 6 | 973 | |||
XM_017023517.1 | XP_016879006.1 | 6 | 704 | |||
XM_017023518.1 | XP_016879007.1 | 6 | 792 | |||
XM_017023519.1 | XP_016879008.1 | 6 | 849 | |||
XM_017023521.1 | XP_016879010.1 | 1 | 99 | |||
GenBank mRNA | BQ433543.1 | - | 561 | |||
DB231720.1 | - | 5 | 507 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | KF175195.1 | - | 4 | 349 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 5 | 521 | ||
GenBank mRNA | AB209632.1 | - | 4 | 3088 | ||
AK294452.1 | - | 1 | 84 | |||
AK295170.1 | - | 5 | 520 | |||
AK303500.1 | - | 1 | 15 | |||
BC036263.1 | - | 5 | 521 | 3903371 | ||
BC094882.1 | - | 4 | 448 | 3093162 | ||
BM557638.1 | - | 1 | 95 | |||
KF175175.1 | - | 4 | 388 | |||
KF175176.1 | - | 4 | 445 | |||
KF175177.1 | - | 4 | 445 | |||
KF175178.1 | - | 4 | 346 | |||
KF175179.1 | - | 4 | 388 | |||
KF175180.1 | - | 4 | 388 | |||
KF175181.1 | - | 4 | 532 | |||
KF175182.1 | - | 4 | 388 | |||
KF175183.1 | - | 3 | 280 | |||
KF175184.1 | - | 3 | 271 | |||
KF175185.1 | - | 4 | 388 | |||
KF175186.1 | - | 3 | 262 | |||
KF175187.1 | - | 4 | 388 | |||
KF175188.1 | - | 4 | 349 | |||
KF175189.1 | - | 4 | 349 | |||
KF175190.1 | - | 4 | 388 | |||
KF175191.1 | - | 4 | 349 | |||
KF175192.1 | - | 4 | 349 | |||
KF175193.1 | - | 4 | 352 | |||
KF175194.1 | - | 4 | 352 | |||
KF175196.1 | - | 4 | 349 | |||
KF175197.1 | - | 4 | 349 | |||
KF175198.1 | - | 4 | 445 | |||
KF175199.1 | - | 4 | 445 | |||
KF175200.1 | - | 4 | 445 | |||
KF175201.1 | - | 4 | 388 | |||
KF175202.1 | - | 4 | 349 | |||
KF175203.1 | - | 4 | 445 | |||
KF175205.1 | - | 4 | 445 | |||
KF175206.1 | - | 4 | 445 | |||
KF175215.1 | - | 6 | 707 | |||
KF175216.1 | - | 5 | 722 | |||
KF175218.1 | - | 4 | 350 | |||
KF175223.1 | - | 4 | 445 | |||
KF175224.1 | - | 4 | 388 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 5 | 670 | ||
XM_017023244.1 | XP_016878733.1 | 4 | 2681 | |||
XM_017023245.1 | XP_016878734.1 | 5 | 2518 | |||
XM_017023246.1 | XP_016878735.1 | 5 | 2439 | |||
XM_017023247.1 | XP_016878736.1 | 4 | 672 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
