siRNA Details
Gene Symbol: NPIPB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135865.1 | NP_001129337.1 | 7 | 3772 | ||
GenBank mRNA | AK296301.1 | - | 3381 |
Gene Symbol: LOC613037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321892.1 | NP_001308821.1 | 9 | 3701 |
Gene Symbol: NPIPB11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310137.1 | NP_001297066.1 | 8 | 3460 |
Gene Symbol: NPIPB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130464.2 | NP_569731.2 | 9 | 3569 | ||
GenBank mRNA | AB209632.1 | - | 11 | 5278 | ||
AK294452.1 | - | 5 | 3132 | |||
AK300277.1 | - | 3264 | ||||
BC094882.1 | - | 8 | 2372 | 3093162 | ||
BQ950195.1 | - | 1 | 540 |
Gene Symbol: NPIPB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310148.1 | NP_001297077.1 | 8 | 3985 | ||
XM_017023244.1 | XP_016878733.1 | 7 | 5996 | |||
XM_017023245.1 | XP_016878734.1 | 8 | 5833 | |||
XM_017023246.1 | XP_016878735.1 | 8 | 5754 | |||
XM_017023247.1 | XP_016878736.1 | 9 | 3741 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
