siRNA Details
Gene Symbol: CHADL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138481.1 | NP_612490.1 | 6 | 2329 | ||
XM_011529933.2 | XP_011528235.1 | 5 | 2991 | |||
GenBank mRNA | BC012882.1 | - | 3 | 376 | ||
BC019839.1 | - | 4 | 1516 | |||
BC040188.1 | - | 6 | 2069 | |||
BC048421.1 | - | 4 | 1352 | |||
BC068590.1 | - | 6 | 2329 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
