siRNA Details
Gene Symbol: ZBTB42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001137601.2 | NP_001131073.1 | 1 | 3204 | ||
XM_017020911.1 | XP_016876400.1 | 2 | 3308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
