siRNA Details
Gene Symbol: MAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030885.3 | NP_112147.2 | 4 | 1394 | ||
GenBank mRNA | AK091042.1 | - | 1 | 586 | ||
BC051843.1 | - | 4 | 939 | |||
DA150735.1 | - | 1 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
