siRNA Details
Gene Symbol: C14orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134877.1 | NP_001128349.1 | 2 | 100 | ||
NM_001198983.1 | NP_001185912.1 | 2 | 176 | |||
GenBank mRNA | BX161477.1 | - | 2 | 176 | ||
BX248038.1 | - | 2 | 100 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
