siRNA Details
Gene Symbol: LHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000894.2 | NP_000885.1 | 3 | 425 | ||
XM_011526975.1 | XP_011525277.1 | 2 | 499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
