siRNA Details
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017940.4 | NP_060410.2 | 3735 | |||
GenBank mRNA | AB051480.1 | - | 15 | 1792 | ||
AF379619.1 | - | 1 | 300 | |||
AF379620.1 | - | 1 | 271 | |||
AF379624.1 | - | 5 | 520 | |||
AF379628.1 | - | 1 | 112 | |||
AY894575.1 | - | 26 | 3617 | |||
BC034418.1 | - | 10 | 1297 | |||
BC094705.1 | - | 8 | 943 |
Gene Symbol: LOC102724250 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546188.2 | XP_011544490.1 | 16 | 10832 |
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 2401 | |||
NM_001302371.1 | NP_001289300.1 | 2401 | ||||
GenBank mRNA | AF379616.1 | - | 1 | 213 | ||
AF379632.1 | - | 3 | 209 | |||
AF379635.1 | - | 10 | 1214 | |||
AF380580.1 | - | 7 | 655 | |||
AF380582.1 | - | 3 | 293 | |||
AF419616.1 | - | 3 | 140 | |||
AK000726.1 | - | 14 | 2465 | |||
AL050141.1 | - | 1 | 869 | |||
AY894565.1 | - | 17 | 2213 | |||
BC021111.1 | - | 15 | 1995 | |||
BC063799.1 | - | 15 | 1978 | |||
BX538005.1 | - | 18 | 2692 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 16 | 2241 | ||
NM_183372.5 | NP_899228.4 | 18 | 2935 | |||
NR_046188.3 | 19 | 3003 | ||||
NR_110864.1 | 17 | 2927 | ||||
GenBank mRNA | AF419617.1 | - | 17 | 2354 | ||
AK054850.1 | - | 2 | 680 | |||
AK302413.1 | - | 15 | 2077 | |||
AL832622.2 | - | 19 | 3138 | |||
BC136292.1 | - | 18 | 2935 | 9020279 | ||
BX648497.1 | - | 17 | 2726 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 25 | 3946 | ||
GenBank mRNA | AB033071.1 | - | 25 | 3419 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 14 | 1916 | ||
XM_011509395.2 | XP_011507697.1 | 17 | 2416 | |||
GenBank mRNA | AF161426.1 | - | 2 | 92 | ||
AF379626.1 | - | 4 | 309 | |||
AF379631.1 | - | 5 | 401 | |||
AK095459.1 | - | 4 | 1101 | |||
AL117237.1 | - | 8 | 913 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 16 | 2265 | ||
NM_173638.4 | NP_775909.2 | 13 | 1735 | |||
XM_017001077.1 | XP_016856566.1 | 11 | 1383 | |||
GenBank mRNA | AF379630.1 | - | 8 | 913 | ||
AF420437.1 | - | 1 | 271 | |||
AK097180.1 | - | 16 | 2184 | |||
AK290142.1 | - | 5 | 437 | |||
AK290302.1 | - | 10 | 1194 | |||
AK299360.1 | - | 7 | 775 | |||
AY894566.1 | - | 10 | 1175 | |||
BC023087.1 | - | 13 | 1592 | 3918932 | ||
DA932476.1 | - | 4 | 319 | |||
DB185273.1 | - | 4 | 326 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 11 | 1376 | ||
GenBank mRNA | AF379633.1 | - | 3 | 226 | ||
AF379636.1 | - | 1 | 26 | |||
AK292666.1 | - | 2 | 129 | |||
AK295639.1 | - | 5 | 847 | |||
CR936830.1 | - | 2 | 130 |
Gene Symbol: NBPF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278267.1 | NP_001265196.1 | 1271 | |||
GenBank mRNA | AK299612.1 | - | 2 | 85 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 13 | 1560 | |||
GenBank mRNA | AK125792.1 | - | 13 | 1560 | ||
DC403040.1 | - | 3 | 156 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 14 | 1946 | ||
XM_017003011.1 | XP_016858500.1 | 14 | 1945 | |||
GenBank mRNA | AK294944.1 | - | 8 | 929 | ||
DA203219.1 | - | 5 | 389 | |||
DA675097.1 | - | 5 | 364 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 14 | 2088 | ||
NR_102404.1 | 13 | 1924 | ||||
NR_102405.1 | 12 | 1821 | ||||
GenBank mRNA | AK300749.1 | - | 5 | 348 | ||
AY894561.1 | - | 11 | 1338 | |||
AY894562.1 | - | 9 | 1113 | |||
AY894563.1 | - | 8 | 987 | |||
AY894567.1 | - | 12 | 1821 | |||
AY894568.1 | - | 13 | 1719 | |||
AY894569.1 | - | 13 | 1721 | |||
AY894570.1 | - | 9 | 1113 | |||
AY894571.1 | - | 13 | 1985 | |||
AY894572.1 | - | 13 | 1924 | |||
AY894573.1 | - | 13 | 1985 | |||
BC086308.1 | - | 14 | 1931 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 15 | 1991 | ||
NM_001277444.1 | NP_001264373.1 | 15 | 1991 | |||
XM_006711317.2 | XP_006711380.1 | 16 | 2219 | |||
GenBank mRNA | AF419618.1 | - | 12 | 1390 | ||
AK095030.1 | - | 2 | 2267 | |||
AK294414.1 | - | 9 | 1149 | |||
AY894574.1 | - | 19 | 2443 | |||
BC071995.1 | - | 10 | 1175 | |||
BC094841.1 | - | 10 | 1175 | |||
BQ436001.1 | - | 3 | 175 | |||
DQ786323.1 | - | 2 | 164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
