siRNA Details
Gene Symbol: NBPF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039703.5 | NP_001034792.4 | 2295 | |||
NM_001302371.1 | NP_001289300.1 | 2295 | ||||
GenBank mRNA | AF379632.1 | - | 5, 6 | 553 | ||
AF380582.1 | - | 1, 2 | 187 | |||
AL050141.1 | - | 4, 5 | 2091 | |||
AY894565.1 | - | 15, 16 | 2107 | |||
BX538005.1 | - | 20, 21 | 3035 |
Gene Symbol: NBPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB051480.1 | - | 17, 18 | 2136 | ||
AF379624.1 | - | 7, 8 | 864 | |||
BC034418.1 | - | 12, 13 | 1641 | |||
BC094705.1 | - | 10, 11 | 1287 |
Gene Symbol: NBPF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101663.4 | NP_001095133.3 | 18, 19 | 2585 | ||
NM_183372.5 | NP_899228.4 | 20, 21 | 3279 | |||
NR_046188.3 | 21, 22 | 3347 | ||||
NR_110864.1 | 19, 20 | 3271 | ||||
GenBank mRNA | AF419617.1 | - | 19, 20 | 2698 | ||
AK054850.1 | - | 4, 5 | 1024 | |||
AK302413.1 | - | 17, 18 | 2421 | |||
AL832622.2 | - | 21, 22 | 3482 | |||
BC136292.1 | - | 20, 21 | 3279 | 9020279 | ||
BX648497.1 | - | 19, 20 | 3070 |
Gene Symbol: NBPF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278141.1 | NP_001265070.1 | 27, 28 | 4290 | ||
GenBank mRNA | AB033071.1 | - | 27, 28 | 3763 |
Gene Symbol: NBPF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015383.2 | NP_056198.2 | 40, 41 | 5188 | ||
XM_011509395.2 | XP_011507697.1 | 43, 44 | 5688 | |||
GenBank mRNA | AF379626.1 | - | 2, 3 | 203 | ||
AK095459.1 | - | 8, 9 | 1727 | |||
AL117237.1 | - | 10, 11 | 1257 |
Gene Symbol: NBPF15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170755.2 | NP_001164226.1 | 18, 19 | 2609 | ||
NM_173638.4 | NP_775909.2 | 15, 16 | 2079 | |||
XM_017001077.1 | XP_016856566.1 | 13, 14 | 1727 | |||
GenBank mRNA | AF379630.1 | - | 10, 11 | 1257 | ||
AK097180.1 | - | 18, 19 | 2528 | |||
AK290142.1 | - | 7, 8 | 781 | |||
AK290302.1 | - | 12, 13 | 1538 | |||
AK299360.1 | - | 9, 10 | 1119 | |||
AY894566.1 | - | 12, 13 | 1519 | |||
BC023087.1 | - | 15, 16 | 1936 | 3918932 | ||
DA932476.1 | - | 2, 3 | 213 |
Gene Symbol: NBPF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017003019.1 | XP_016858508.1 | 13, 14 | 1720 | ||
GenBank mRNA | AF379633.1 | - | 1, 2 | 120 | ||
AF379636.1 | - | 5, 6 | 652 | |||
AK292666.1 | - | 4, 5 | 473 |
Gene Symbol: NBPF25P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104217.1 | 15, 16 | 1904 | |||
GenBank mRNA | AK125792.1 | - | 15, 16 | 1904 |
Gene Symbol: NBPF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542529.2 | XP_011540831.1 | 16, 17 | 2290 | ||
XM_017003011.1 | XP_016858500.1 | 16, 17 | 2289 | |||
GenBank mRNA | AK294944.1 | - | 19, 20 | 2173 | ||
DA203219.1 | - | 3, 4 | 283 | |||
DA675097.1 | - | 3, 4 | 258 |
Gene Symbol: NBPF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037501.2 | NP_001032590.2 | 16, 17 | 2432 | ||
NR_102404.1 | 15, 16 | 2268 | ||||
NR_102405.1 | 14, 15 | 2165 | ||||
GenBank mRNA | AY894561.1 | - | 9, 10 | 1232 | ||
AY894562.1 | - | 11, 12 | 1457 | |||
AY894563.1 | - | 10, 11 | 1331 | |||
AY894567.1 | - | 14, 15 | 2165 | |||
AY894568.1 | - | 15, 16 | 2063 | |||
AY894569.1 | - | 15, 16 | 2065 | |||
AY894570.1 | - | 11, 12 | 1457 | |||
AY894571.1 | - | 15, 16 | 2329 | |||
AY894572.1 | - | 15, 16 | 2268 | |||
AY894573.1 | - | 15, 16 | 2329 | |||
BC086308.1 | - | 16, 17 | 2275 | 5453529 |
Gene Symbol: NBPF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037675.3 | NP_001032764.2 | 17, 18 | 2335 | ||
NM_001277444.1 | NP_001264373.1 | 17, 18 | 2335 | |||
XM_006711317.2 | XP_006711380.1 | 18, 19 | 2563 | |||
GenBank mRNA | AF419618.1 | - | 10, 11 | 1284 | ||
AK095030.1 | - | 4, 5 | 2611 | |||
AK294414.1 | - | 11, 12 | 1493 | |||
AY894574.1 | - | 17, 18 | 2337 | |||
BC071995.1 | - | 12, 13 | 1519 | |||
BC094841.1 | - | 12, 13 | 1519 | |||
BQ436001.1 | - | 1, 2 | 69 | |||
DQ786323.1 | - | 4, 5 | 508 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
