siRNA Details
Gene Symbol: SMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017411.3 | NP_059107.1 | 9 | 1355 | ||
NM_022875.2 | NP_075013.1 | 8 | 1301 | |||
NM_022876.2 | NP_075014.1 | 8 | 1259 | |||
NM_022877.2 | NP_075015.1 | 7 | 1205 | |||
XM_011543600.2 | XP_011541902.1 | 8 | 1039 | |||
XM_011543601.1 | XP_011541903.1 | 7 | 985 | |||
XM_011543602.2 | XP_011541904.1 | 7 | 955 | |||
XM_011543603.2 | XP_011541905.1 | 6 | 901 | |||
GenBank mRNA | BC000908.2 | - | 8 | 1170 | 2985046 | |
BC015308.1 | - | 9 | 1222 | 4427000 | ||
BC070242.1 | - | 8 | 1147 | 4673318 | ||
DQ185039.1 | - | 1 | 411 | |||
JQ690861.1 | - | 6 | 743 | |||
JQ690862.1 | - | 5 | 689 | |||
JQ690863.1 | - | 4 | 593 | |||
JQ690864.1 | - | 5 | 737 | |||
JQ690865.1 | - | 5 | 794 | |||
JQ690866.1 | - | 5 | 647 |
Gene Symbol: SMN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000344.3 | NP_000335.1 | 9 | 1355 | ||
NM_001297715.1 | NP_001284644.1 | 8 | 1301 | |||
NM_022874.2 | NP_075012.1 | 8 | 1259 | |||
XM_011543597.1 | XP_011541899.1 | 8 | 1039 | |||
XM_011543598.2 | XP_011541900.1 | 7 | 954 | |||
XM_017009786.1 | XP_016865275.1 | 7 | 1084 | |||
GenBank mRNA | BC062723.1 | - | 9 | 1211 | 4250429 | |
JQ690867.1 | - | 5 | 689 | |||
JQ690868.1 | - | 4 | 593 | |||
JQ745297.1 | - | 5 | 590 | |||
U18423.1 | - | 9 | 1223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
