siRNA Details
Gene Symbol: SMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017411.3 | NP_059107.1 | 7, 8 | 1006 | ||
NM_022876.2 | NP_075014.1 | 6, 7 | 910 | |||
XM_011543600.2 | XP_011541902.1 | 6, 7 | 690 | |||
XM_011543602.2 | XP_011541904.1 | 5, 6 | 606 | |||
GenBank mRNA | BC015308.1 | - | 7, 8 | 873 | 4427000 | |
JQ690861.1 | - | 4, 5 | 394 | |||
JQ690866.1 | - | 3, 4 | 298 | |||
U21914.1 | - | 7, 8 | 840 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
