siRNA Details
Gene Symbol: ATXN7L3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136262.1 | NP_001129734.1 | 1 | 2551 | ||
GenBank mRNA | AF456479.1 | - | 1 | 1033 | ||
AK054742.1 | - | 1 | 542 | |||
AK056642.1 | - | 1 | 2492 | |||
AK095526.1 | - | 1 | 1432 | |||
AL713650.2 | - | 1 | 2445 | |||
BC009111.1 | - | 1 | 2454 | 3460701 | ||
BX647460.2 | - | 1 | 2443 | |||
CX868912.1 | - | 1 | 620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
