siRNA Details
Gene Symbol: ATXN7L3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136262.1 | NP_001129734.1 | 1 | 374 | ||
GenBank mRNA | AK056642.1 | - | 1 | 318 | ||
AL713650.2 | - | 1 | 267 | |||
BC009111.1 | - | 1 | 278 | 3460701 | ||
BX647460.2 | - | 1 | 267 | |||
DA298752.1 | - | 1 | 374 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
